Assay Details
| Gene Symbol: nsun4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008649.1 | NP_001008649.1 | 4 - 5 | 796 | 139 | |
| XM_021480053.1 | XP_021335728.1 | 4 - 5 | 822 | 139 | ||
| GenBank mRNA | BC086725.1 | - | 4 - 5 | 796 | 139 | |
More Information
Set Membership: |
|

nsun4
Genomic Map