Assay Details
| Gene Symbol: gh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020492.2 | NP_001018328.2 | 3 - 4 | 316 | 68 | |
| GenBank mRNA | AJ937858.1 | - | 3 - 4 | 308 | 68 | |
| AY286447.1 | - | 1 - 2 | 105 | 68 | ||
| BC116501.1 | - | 3 - 4 | 316 | 68 | ||
More Information
Set Membership: |
|

gh1
Genomic Map