Assay Details
| Gene Symbol: neurl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001326539.1 | NP_001313468.1 | 4 - 5 | 1706 | 110 | |
| XM_005157198.4 | XP_005157255.1 | 4 - 5 | 1965 | 110 | ||
| GenBank mRNA | BC095256.1 | - | 4 - 5 | 1184 | 110 | |
More Information
Set Membership: |
|

neurl1b
Genomic Map