Assay Details
| Gene Symbol: sptlc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020619.1 | NP_001018455.1 | 7 - 8 | 1034 | 79 | |
| GenBank mRNA | BC056779.1 | - | 8 - 9 | 1177 | 79 | |
| BC095355.1 | - | 7 - 8 | 1034 | 79 | ||
| BC165370.1 | - | 7 - 8 | 904 | 79 | ||
More Information
Set Membership: |
|

sptlc2a
Genomic Map