Assay Details
| Gene Symbol: kcnj8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030153.1 | NP_001025324.1 | 1 - 2 | 344 | 85 | |
| GenBank mRNA | AB231936.1 | - | 1 - 2 | 344 | 85 | |
| BC163413.1 | - | 1 - 2 | 415 | 85 | ||
| BC163434.1 | - | 1 - 2 | 415 | 85 | ||
More Information
Set Membership: |
|

kcnj8
Genomic Map