Assay Details
| Gene Symbol: tgfbr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037683.2 | NP_001032772.2 | 7 - 8 | 1264 | 67 | |
| GenBank mRNA | BC109402.1 | - | 7 - 8 | 1273 | 67 | |
| EF462417.1 | - | 7 - 8 | 1264 | 67 | ||
More Information
Set Membership: |
|

tgfbr1a
Genomic Map