Assay Details
| Gene Symbol: ripk1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043350.1 | NP_001036815.1 | 6 - 7 | 877 | 117 | |
| GenBank mRNA | DQ848680.1 | - | 6 - 7 | 877 | 117 | |
More Information
Set Membership: |
|

ripk1l
Genomic Map