Assay Details
| Gene Symbol: atoh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079991.2 | NP_001073460.2 | 1 - 2 | 880 | 66 | |
| GenBank mRNA | BC128838.1 | - | 1 - 2 | 837 | 66 | |
| EU272033.1 | - | 1 - 2 | 632 | 66 | ||
More Information
Set Membership: |
|

atoh8
Genomic Map