Assay Details
| Gene Symbol: atoh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079991.2 | NP_001073460.2 | 2 - 3 | 1071 | 88 | |
| GenBank mRNA | BC128838.1 | - | 2 - 3 | 1028 | 88 | |
More Information
Set Membership: |
|

atoh8
Genomic Map