Assay Details
| Gene Symbol: cntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080051.1 | NP_001073520.1 | 12 - 13 | 1703 | 86 | |
| GenBank mRNA | BC124394.1 | - | 12 - 13 | 1703 | 86 | |
More Information
Set Membership: |
|

cntn4
Genomic Map