Assay Details
| Gene Symbol: srpk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080669.1 | NP_001074138.1 | 10 - 11 | 1036 | 82 | |
| XM_005155658.4 | XP_005155715.1 | 9 - 10 | 1056 | 82 | ||
| XM_009305766.3 | XP_009304041.1 | 9 - 10 | 1770 | 82 | ||
| XM_009305767.2 | XP_009304042.1 | 10 - 11 | 1048 | 82 | ||
| GenBank mRNA | BC129460.1 | - | 10 - 11 | 1036 | 82 | |
More Information
Set Membership: |
|

srpk1b
Genomic Map