Assay Details
| Gene Symbol: dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089397.1 | NP_001082866.1 | 5 - 6 | 1198 | 73 | |
| GenBank mRNA | BC134939.1 | - | 5 - 6 | 1198 | 73 | |
More Information
Set Membership: |
|

dph2
Genomic Map