Assay Details
| Gene Symbol: ubtd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089401.1 | NP_001082870.1 | 1 - 2 | 382 | 81 | |
| GenBank mRNA | BC139662.1 | - | 1 - 2 | 382 | 81 | |
More Information
Set Membership: |
|

ubtd1a
Genomic Map