Assay Details
| Gene Symbol: rock2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093747.2 | NP_001087216.1 | 18 - 19 | 2327 | 59 | |
| XM_009294695.3 | XP_009292970.1 | 18 - 19 | 2799 | 59 | ||
| XM_009294697.3 | XP_009292972.1 | 6 - 7 | 865 | 59 | ||
| XM_017352433.2 | XP_017207922.1 | 18 - 19 | 2800 | 59 | ||
| XM_017352434.2 | XP_017207923.1 | 18 - 19 | 2800 | 59 | ||
| XM_017352437.2 | XP_017207926.1 | 18 - 19 | 2803 | 59 | ||
| XM_021468356.1 | XP_021324031.1 | 18 - 19 | 2802 | 59 | ||
| XM_021468357.1 | XP_021324032.1 | 18 - 19 | 2801 | 59 | ||
| XM_021468358.1 | XP_021324033.1 | 18 - 19 | 2801 | 59 | ||
| XM_021468359.1 | XP_021324034.1 | 18 - 19 | 2801 | 59 | ||
| XM_021468360.1 | XP_021324035.1 | 18 - 19 | 2804 | 59 | ||
| XM_021468361.1 | XP_021324036.1 | 18 - 19 | 2802 | 59 | ||
More Information
Set Membership: |
|

rock2b
Genomic Map