Assay Details
| Gene Symbol: rock2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093747.2 | NP_001087216.1 | 28 - 29 | 3581 | 104 | |
| GenBank mRNA | GQ395807.1 | - | 28 - 29 | 3581 | 104 | |
More Information
Set Membership: |
|

rock2b
Genomic Map