Assay Details
| Gene Symbol: rock2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093747.2 | NP_001087216.1 | 30 - 31 | 3922 | 75 | |
| XM_009294695.3 | XP_009292970.1 | 30 - 31 | 4394 | 75 | ||
| XM_009294697.3 | XP_009292972.1 | 19 - 20 | 2661 | 75 | ||
| XM_017352433.2 | XP_017207922.1 | 31 - 32 | 4596 | 75 | ||
| XM_017352434.2 | XP_017207923.1 | 30 - 31 | 4395 | 75 | ||
| XM_021468356.1 | XP_021324031.1 | 31 - 32 | 4598 | 75 | ||
| XM_021468357.1 | XP_021324032.1 | 31 - 32 | 4597 | 75 | ||
| XM_021468358.1 | XP_021324033.1 | 31 - 32 | 4597 | 75 | ||
| XM_021468359.1 | XP_021324034.1 | 31 - 32 | 4597 | 75 | ||
| GenBank mRNA | GQ395807.1 | - | 30 - 31 | 3922 | 75 | |
| Gene Symbol: LOC101884327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009304741.3 | XP_009303016.1 | 1 - 2 | 243 | 75 | |
| XM_021470959.1 | XP_021326634.1 | 1 - 2 | 229 | 75 | ||
| Gene Symbol: LOC110438182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021468645.1 | XP_021324320.1 | 1 - 2 | 244 | 75 | |
| XM_021468646.1 | XP_021324321.1 | 1 - 2 | 229 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009304741.3, XM_021468645.1, XM_021468646.1, XM_021470959.1 transcripts.
Set Membership: |
|

rock2b
Genomic Map