Assay Details
| Gene Symbol: rock2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093747.2 | NP_001087216.1 | 3 - 4 | 287 | 78 | |
| XM_009294695.3 | XP_009292970.1 | 3 - 4 | 759 | 78 | ||
| XM_017352433.2 | XP_017207922.1 | 3 - 4 | 760 | 78 | ||
| XM_017352434.2 | XP_017207923.1 | 3 - 4 | 760 | 78 | ||
| XM_017352437.2 | XP_017207926.1 | 3 - 4 | 763 | 78 | ||
| XM_021468356.1 | XP_021324031.1 | 3 - 4 | 762 | 78 | ||
| XM_021468357.1 | XP_021324032.1 | 3 - 4 | 761 | 78 | ||
| XM_021468358.1 | XP_021324033.1 | 3 - 4 | 761 | 78 | ||
| XM_021468359.1 | XP_021324034.1 | 3 - 4 | 761 | 78 | ||
| XM_021468360.1 | XP_021324035.1 | 3 - 4 | 764 | 78 | ||
| XM_021468361.1 | XP_021324036.1 | 3 - 4 | 762 | 78 | ||
| GenBank mRNA | GQ395807.1 | - | 3 - 4 | 287 | 78 | |
| Gene Symbol: LOC108182331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017354463.2 | XP_017209952.1 | 1 - 2 | 550 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017354463.2 transcripts.
Set Membership: |
|

rock2b
Genomic Map