Assay Details
| Gene Symbol: supt16h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097584.2 | NP_001091053.1 | 19 - 20 | 2764 | 55 | |
| GenBank mRNA | BC163334.1 | - | 19 - 20 | 2526 | 55 | |
| BC163336.1 | - | 19 - 20 | 2526 | 55 | ||
| EF527261.1 | - | 19 - 20 | 2490 | 55 | ||
More Information
Set Membership: |
|

supt16h
Genomic Map