Assay Details
| Gene Symbol: borcs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098247.2 | NP_001091717.1 | 3 - 4 | 257 | 94 | |
| XM_005170457.4 | XP_005170514.1 | 3 - 4 | 331 | 94 | ||
| GenBank mRNA | BC162286.1 | - | 3 - 4 | 257 | 94 | |
| BC162309.1 | - | 3 - 4 | 257 | 94 | ||
More Information
Set Membership: |
|

borcs8
Genomic Map