Assay Details
| Gene Symbol: borcs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098247.2 | NP_001091717.1 | 5 - 6 | 435 | 81 | |
| GenBank mRNA | BC162286.1 | - | 5 - 6 | 435 | 81 | |
| BC162309.1 | - | 5 - 6 | 435 | 81 | ||
More Information
Set Membership: |
|

borcs8
Genomic Map