Assay Details
| Gene Symbol: jam2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098264.1 | NP_001091734.1 | 5 - 6 | 761 | 50 | |
| GenBank mRNA | CU458944.1 | - | 5 - 6 | 761 | 50 | |
More Information
Set Membership: |
|

jam2a
Genomic Map