Assay Details
| Gene Symbol: rims3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100009.2 | NP_001093479.1 | 1 - 2 | 206 | 72 | |
| XM_005170052.4 | XP_005170109.1 | 2 - 3 | 484 | 72 | ||
More Information
Set Membership: |
|

rims3
Genomic Map