Assay Details
| Gene Symbol: frzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130943.2 | NP_571018.1 | 1 - 2 | 775 | 97 | |
| GenBank mRNA | AF116853.1 | - | 1 - 2 | 459 | 97 | |
| BC162928.1 | - | 1 - 2 | 527 | 97 | ||
| BC162950.1 | - | 1 - 2 | 527 | 97 | ||
| CT663963.2 | - | 1 - 2 | 227 | 97 | ||
| KM655689.1 | - | 1 - 2 | 459 | 97 | ||
More Information
Set Membership: |
|

frzb
Genomic Map