Assay Details
| Gene Symbol: psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131024.1 | NP_571099.1 | 2 - 3 | 368 | 152 | |
| XM_005169882.4 | XP_005169939.1 | 3 - 4 | 457 | 152 | ||
| XM_005169883.4 | XP_005169940.1 | 3 - 4 | 458 | 152 | ||
| GenBank mRNA | AJ132931.1 | - | 2 - 3 | 384 | 152 | |
| BC054639.1 | - | 3 - 4 | 381 | 152 | ||
More Information
Set Membership: |
|

psen1
Genomic Map