Assay Details
| Gene Symbol: psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131024.1 | NP_571099.1 | 6 - 7 | 895 | 128 | |
| XM_005169882.4 | XP_005169939.1 | 7 - 8 | 984 | 128 | ||
| XM_005169883.4 | XP_005169940.1 | 7 - 8 | 985 | 128 | ||
| GenBank mRNA | AJ132931.1 | - | 6 - 7 | 911 | 128 | |
More Information
Set Membership: |
|

psen1
Genomic Map