Assay Details
| Gene Symbol: psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131024.1 | NP_571099.1 | 7 - 8 | 986 | 111 | |
| XM_005169882.4 | XP_005169939.1 | 8 - 9 | 1075 | 111 | ||
| GenBank mRNA | AJ132931.1 | - | 7 - 8 | 1002 | 111 | |
More Information
Set Membership: |
|

psen1
Genomic Map