Assay Details
| Gene Symbol: psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131024.1 | NP_571099.1 | 8 - 9 | 1157 | 59 | |
| XM_005169882.4 | XP_005169939.1 | 9 - 10 | 1246 | 59 | ||
| XM_005169883.4 | XP_005169940.1 | 9 - 10 | 1244 | 59 | ||
| GenBank mRNA | AJ132931.1 | - | 8 - 9 | 1173 | 59 | |
| BC054639.1 | - | 9 - 10 | 1167 | 59 | ||
More Information
Set Membership: |
|

psen1
Genomic Map