Assay Details
| Gene Symbol: psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131024.1 | NP_571099.1 | 9 - 10 | 1270 | 99 | |
| XM_005169882.4 | XP_005169939.1 | 10 - 11 | 1359 | 99 | ||
| XM_005169883.4 | XP_005169940.1 | 10 - 11 | 1357 | 99 | ||
| GenBank mRNA | AJ132931.1 | - | 9 - 10 | 1286 | 99 | |
| BC054639.1 | - | 10 - 11 | 1280 | 99 | ||
More Information
Set Membership: |
|

psen1
Genomic Map