Assay Details
| Gene Symbol: thraa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131396.1 | NP_571471.1 | 2 - 3 | 412 | 95 | |
| XM_005163986.4 | XP_005164043.1 | 2 - 3 | 1017 | 95 | ||
| XM_021469328.1 | XP_021325003.1 | 2 - 3 | 235 | 95 | ||
| GenBank mRNA | BC096778.1 | - | 2 - 3 | 412 | 95 | |
| BC164729.1 | - | 1 - 2 | 81 | 95 | ||
| U54796.1 | - | 2 - 3 | 412 | 95 | ||
More Information
Set Membership: |
|

thraa
Genomic Map