Assay Details
| Gene Symbol: thraa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131396.1 | NP_571471.1 | 6 - 7 | 950 | 90 | |
| XM_005163985.4 | XP_005164042.1 | 5 - 6 | 678 | 90 | ||
| XM_005163986.4 | XP_005164043.1 | 6 - 7 | 1555 | 90 | ||
| XM_009299516.3 | XP_009297791.1 | 5 - 6 | 678 | 90 | ||
| XM_021469328.1 | XP_021325003.1 | 6 - 7 | 773 | 90 | ||
| GenBank mRNA | BC096778.1 | - | 6 - 7 | 950 | 90 | |
| BC164729.1 | - | 5 - 6 | 619 | 90 | ||
| U54796.1 | - | 6 - 7 | 950 | 90 | ||
More Information
Set Membership: |
|

thraa
Genomic Map