Assay Details
| Gene Symbol: atp5mc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131761.1 | NP_571836.1 | 2 - 3 | 342 | 97 | |
| XM_005165783.4 | XP_005165840.1 | 3 - 4 | 264 | 97 | ||
| GenBank mRNA | BC045894.1 | - | 2 - 3 | 342 | 97 | |
More Information
Set Membership: |
|

atp5mc3b
Genomic Map