Assay Details
| Gene Symbol: fgfr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309399.1 | NP_001296328.1 | 10 - 11 | 2034 | 99 | |
| NM_001309400.1 | NP_001296329.1 | 8 - 9 | 1698 | 99 | ||
| NM_152962.3 | NP_694494.2 | 10 - 11 | 2040 | 99 | ||
| XM_009304303.3 | XP_009302578.1 | 10 - 11 | 2085 | 99 | ||
| XM_009304304.3 | XP_009302579.1 | 10 - 11 | 2080 | 99 | ||
| GenBank mRNA | AF389400.2 | - | 9 - 10 | 1422 | 99 | |
| AY197498.1 | - | 9 - 10 | 1416 | 99 | ||
| AY197499.1 | - | 8 - 9 | 1272 | 99 | ||
| AY197500.1 | - | 7 - 8 | 1080 | 99 | ||
| BC162342.1 | - | 9 - 10 | 1422 | 99 | ||
| CU458752.1 | - | 9 - 10 | 1375 | 99 | ||
More Information
Set Membership: |
|

fgfr1a
Genomic Map