Assay Details
| Gene Symbol: pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153654.2 | NP_705940.1 | 10 - 11 | 1264 | 57 | |
| XM_009305360.3 | XP_009303635.1 | 11 - 12 | 1346 | 57 | ||
| GenBank mRNA | AB086818.1 | - | 9 - 10 | 1181 | 57 | |
| BC117639.1 | - | 10 - 11 | 1264 | 57 | ||
More Information
Set Membership: |
|

pknox2
Genomic Map