Assay Details
| Gene Symbol: serhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199615.2 | NP_955909.2 | 11 - 12 | 879 | 104 | |
| GenBank mRNA | BC044494.1 | - | 11 - 12 | 879 | 104 | |
| BC066637.1 | - | 11 - 12 | 812 | 104 | ||
| EH483060.1 | - | 2 - 3 | 701 | 104 | ||
More Information
Set Membership: |
|

serhl
Genomic Map