Assay Details
| Gene Symbol: serhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199615.2 | NP_955909.2 | 3 - 4 | 329 | 74 | |
| GenBank mRNA | BC044494.1 | - | 3 - 4 | 329 | 74 | |
| BC066637.1 | - | 3 - 4 | 262 | 74 | ||
| EH457337.1 | - | 3 - 4 | 311 | 74 | ||
More Information
Set Membership: |
|

serhl
Genomic Map