Assay Details
| Gene Symbol: upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200954.1 | NP_957248.1 | 8 - 9 | 1018 | 64 | |
| GenBank mRNA | BC055632.1 | - | 8 - 9 | 1018 | 64 | |
| FM986820.1 | - | 8 - 9 | 1018 | 64 | ||
More Information
Set Membership: |
|

upf3b
Genomic Map