Assay Details
| Gene Symbol: tmem88a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205670.1 | NP_991233.1 | 2 - 3 | 573 | 82 | |
| GenBank mRNA | BC065632.1 | - | 2 - 3 | 573 | 82 | |
| BC152199.1 | - | 2 - 3 | 705 | 82 | ||
| BC155103.1 | - | 2 - 3 | 636 | 82 | ||
More Information
Set Membership: |
|

tmem88a
Genomic Map