Assay Details
| Gene Symbol: tal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213237.1 | NP_998402.1 | 2 - 3 | 597 | 133 | |
| GenBank mRNA | AF038873.1 | - | 1 - 2 | 473 | 133 | |
| BC068324.1 | - | 2 - 3 | 597 | 133 | ||
| EF488003.1 | - | 1 - 2 | 174 | 133 | ||
| EF488004.1 | - | 2 - 3 | 612 | 133 | ||
More Information
Set Membership: |
|

tal1
Genomic Map