Assay Details
| Gene Symbol: cpa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002217.1 | NP_001002217.1 | 8 - 9 | 875 | 72 | |
| GenBank mRNA | BC074071.1 | - | 8 - 9 | 875 | 72 | |
| BC164009.1 | - | 8 - 9 | 816 | 72 | ||
More Information
Set Membership: |
|

cpa4
Genomic Map