Assay Details
| Gene Symbol: aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002423.1 | NP_001002423.1 | 5 - 6 | 655 | 61 | |
| GenBank mRNA | BC076147.1 | - | 5 - 6 | 655 | 61 | |
More Information
Set Membership: |
|

aagab
Genomic Map