Assay Details
| Gene Symbol: mmadhca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002453.1 | NP_001002453.1 | 7 - 8 | 748 | 85 | |
| XM_021469315.1 | XP_021324990.1 | 7 - 8 | 858 | 85 | ||
| GenBank mRNA | BC076018.1 | - | 7 - 8 | 748 | 85 | |
More Information
Set Membership: |
|

mmadhca
Genomic Map