Assay Details
| Gene Symbol: gtf3aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003866.1 | NP_001003866.1 | 8 - 9 | 949 | 68 | |
| GenBank mRNA | AY648780.1 | - | 8 - 9 | 949 | 68 | |
| BC095553.1 | - | 8 - 9 | 947 | 68 | ||
| BC154155.1 | - | 8 - 9 | 872 | 68 | ||
More Information
Set Membership: |
|

gtf3aa
Genomic Map