Assay Details
| Gene Symbol: rcan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005392.1 | NP_001005392.1 | 4 - 5 | 652 | 138 | |
| XM_005158490.3 | XP_005158547.1 | 3 - 4 | 620 | 138 | ||
| GenBank mRNA | BC076514.1 | - | 4 - 5 | 652 | 138 | |
| BC090735.1 | - | 3 - 4 | 613 | 138 | ||
More Information
Set Membership: |
|

rcan3
Genomic Map