Assay Details
| Gene Symbol: aimp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008592.1 | NP_001008592.1 | 6 - 7 | 899 | 116 | |
| GenBank mRNA | BC086748.1 | - | 6 - 7 | 899 | 116 | |
| BC165549.1 | - | 5 - 6 | 711 | 116 | ||
More Information
Set Membership: |
|

aimp1b
Genomic Map