Assay Details
| Gene Symbol: tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316693.1 | NP_001303622.1 | 2 - 3 | 344 | 75 | |
| XM_017352472.2 | XP_017207961.1 | 2 - 3 | 467 | 75 | ||
| GenBank mRNA | BC065982.1 | - | 2 - 3 | 344 | 75 | |
More Information
Set Membership: |
|

tab2
Genomic Map