Assay Details
| Gene Symbol: ankha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030259.2 | NP_001025430.2 | 10 - 11 | 1568 | 71 | |
| XM_021470216.1 | XP_021325891.1 | 8 - 9 | 1246 | 71 | ||
| GenBank mRNA | BC096801.1 | - | 10 - 11 | 71 | ||
More Information
Set Membership: |
|

ankha
Genomic Map