Assay Details
| Gene Symbol: grk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034181.2 | NP_001029353.2 | 6 - 7 | 1517 | 80 | |
| GenBank mRNA | AB212993.1 | - | 6 - 7 | 1517 | 80 | |
| AY900002.1 | - | 6 - 7 | 1461 | 80 | ||
| BC163571.1 | - | 6 - 7 | 1476 | 80 | ||
| BC163572.1 | - | 6 - 7 | 1476 | 80 | ||
More Information
Set Membership: |
|

grk1a
Genomic Map