Assay Details
| Gene Symbol: slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037385.1 | NP_001032462.1 | 9 - 10 | 1201 | 67 | |
| GenBank mRNA | BC109451.1 | - | 9 - 10 | 1201 | 67 | |
More Information
Set Membership: |
|

slc22a18
Genomic Map