Assay Details
| Gene Symbol: dab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040685.1 | NP_001035775.1 | 5 - 6 | 714 | 60 | |
| XM_005170145.4 | XP_005170202.1 | 5 - 6 | 1191 | 60 | ||
| XM_005170147.4 | XP_005170204.1 | 5 - 6 | 1189 | 60 | ||
| XM_005170148.4 | XP_005170205.1 | 5 - 6 | 712 | 60 | ||
| XM_009294566.3 | XP_009292841.1 | 5 - 6 | 1193 | 60 | ||
| XM_009294567.3 | XP_009292842.1 | 5 - 6 | 1192 | 60 | ||
| XM_009294568.3 | XP_009292843.1 | 5 - 6 | 1195 | 60 | ||
| XM_009294569.3 | XP_009292844.1 | 5 - 6 | 1194 | 60 | ||
| XM_021468517.1 | XP_021324192.1 | 5 - 6 | 690 | 60 | ||
| GenBank mRNA | AB690577.1 | - | 4 - 5 | 432 | 60 | |
| BC162673.1 | - | 4 - 5 | 467 | 60 | ||
| DQ166810.1 | - | 5 - 6 | 714 | 60 | ||
| DQ166811.1 | - | 5 - 6 | 714 | 60 | ||
More Information
Set Membership: |
|

dab1a
Genomic Map