Assay Details
| Gene Symbol: dab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040685.1 | NP_001035775.1 | 7 - 8 | 878 | 62 | |
| XM_005170145.4 | XP_005170202.1 | 7 - 8 | 1355 | 62 | ||
| XM_005170147.4 | XP_005170204.1 | 7 - 8 | 1353 | 62 | ||
| XM_009294566.3 | XP_009292841.1 | 7 - 8 | 1357 | 62 | ||
| XM_009294567.3 | XP_009292842.1 | 7 - 8 | 1356 | 62 | ||
| XM_009294568.3 | XP_009292843.1 | 7 - 8 | 1359 | 62 | ||
| XM_021468517.1 | XP_021324192.1 | 7 - 8 | 854 | 62 | ||
| GenBank mRNA | AB690577.1 | - | 6 - 7 | 596 | 62 | |
| BC162673.1 | - | 6 - 7 | 631 | 62 | ||
| DQ166811.1 | - | 7 - 8 | 878 | 62 | ||
More Information
Set Membership: |
|

dab1a
Genomic Map